Please login to your online account to display your discounted pricing

Alfa Aesar™ Catecholborane, 97%

$100.05 - $253.99

Chemical Identifiers

Chemical Name or Material Catecholborane
CAS 274-07-7
Molecular Formula C6H5BO2
Formula Weight 119.92
MDL Number MFCD00005846
UN Number UN2924
Beilstein 972072
Synonym 1,3,2-Benzodioxaborole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1499806

 
alfa aesar™
L1499806
5g Each for $100.05

AAL1499814

 
alfa aesar™
L1499814*
25g Each for $253.99
Description & Specifications

Specifications

Formula Weight 119.92
Chemical Name or Material Catecholborane
TSCA TSCA
Assay 97%
Molecular Formula C6H5BO2
MDL Number MFCD00005846
CAS 274-07-7
UN Number UN2924
Beilstein 972072
Synonym 1,3,2-Benzodioxaborole
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Flash Point 2°C (35°F)
Boiling Point 50°C (50mmHg)
Melting Point 12°C
Quantity 5g

Applications
Catecholborane is used to prepare B-alkylcatecholboranes. It finds application for the preparation of amides and macrocyclic lactams from carboxylic acids. It is used as a stereoselective reducing agent to convert beta-hydroxy ketones to syn 1,3-diols. Further, it reacts with alkyne through hydroboration to form trans vinyl borane, which is a precursor to Suzuki reaction.

Solubility
Miscible with diethyl ether, tetrahydrofuran, dichloromethane, chloroform, carbon tetrachloride, toluene, and benzene.

Notes
Moisture sensitive. Store in a cool place.

Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C