Please login to your online account to display your discounted pricing

Alfa Aesar™ Catecholborane, 50% w/w in toluene (98% dry wt.)

$227.53 - $617.77

Chemical Identifiers

Chemical Name or Material Catecholborane
CAS 274-07-7
Molecular Formula C6H5BO2
Formula Weight 119.92
MDL Number MFCD00005846
UN Number UN2924
Synonym 1,3,2-Benzodioxaborole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3023414

  SDS
alfa aesar™
H3023414
25g Each for $227.53

AAH3023422

  SDS
alfa aesar™
H3023422
100g Each for $617.77
Description & Specifications

Specifications

Formula Weight 119.92
CAS 274-07-7
Concentration 50% w/w in toluene
Synonym 1,3,2-Benzodioxaborole
TSCA TSCA
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Chemical Name or Material Catecholborane
Molecular Formula C6H5BO2
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H225-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Quantity 25g
UN Number UN2924
MDL Number MFCD00005846
Assay Percent Notes (dry wt.)
Assay 98%

Applications
Employed in organic synthesis. Addition of catecholborane to vinylarenes in the presence of Wilkinson′s catalyst, gives Markovnikov hydroboration products exclusively, whereas the corresponding reaction with a-substituted vinylarenesgivesgood yields of vinylboronate. Hydroboration with catecholborane, followed by treatment with easily available reagents such as alkenyl sulfones or alkynyl phenyl sulfones in the presence of a radical initiator, represents an effective and simple one-pot procedure for direct vinylation, formylation, and cyanation. Catecholborane may be used as a stereoselective reducing agent when converting β-hydroxy ketones to syn 1,3-diols.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from water, moisture.

Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.