Please login to your online account to display your discounted pricing

Alfa Aesar™ Catechol-O,O-diacetic acid, 97%

$56.32 - $189.34

Chemical Identifiers

Chemical Name or Material Catechol-O,O-diacetic acid
CAS 5411-14-3
Molecular Formula C10H10O6
Formula Weight 226.18
MDL Number MFCD00014353
Beilstein 2057228
Synonym 1,2-Phenylenedioxydiacetic acid; Pyrocatechol-O,O-diacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0184309

 
alfa aesar™
L0184309
10g Each for $56.32

AAL0184318

 
alfa aesar™
L0184318
50g Each for $189.34
Description & Specifications

Specifications

Formula Weight 226.18
Chemical Name or Material Catechol-O,O-diacetic acid
Assay 97%
Molecular Formula C10H10O6
MDL Number MFCD00014353
CAS 5411-14-3
Synonym 1,2-Phenylenedioxydiacetic acid; Pyrocatechol-O,O-diacetic acid
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 177° to 181°C
Beilstein 2057228
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.