Learn More
Catechol, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 158980050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Catechol | |
103.0°C to 106.0°C | |
245.0°C | |
Authentic | |
Glass bottle | |
MFCD00002188 | |
15,8111 | |
Solubility in water: 430g/L (20°C). Other solubilities: soluble in alcohol, benzene, chloroform, ether, and pyridine and aqueous solutions | |
OC1=CC=CC=C1O | |
110.11 | |
2.38 mm2/s (110°C) | |
110.11 | |
Crystalline Flakes |
120-80-9 | |
Brown-Gray to White | |
131°C | |
99% min. (GC) | |
C6H6O2 | |
5 g | |
pyrocatechol, catechol, 1,2-dihydroxybenzene, 1,2-benzenediol, pyrocatechin, 2-hydroxyphenol, o-benzenediol, pyrocatechine, o-dihydroxybenzene, o-dioxybenzene | |
YCIMNLLNPGFGHC-UHFFFAOYSA-N | |
benzene-1,2-diol | |
289 | |
CHEBI:18135 | |
99+% |
Chemical Identifiers
120-80-9 | |
110.11 | |
YCIMNLLNPGFGHC-UHFFFAOYSA-N | |
289 | |
benzene-1,2-diol |
C6H6O2 | |
MFCD00002188 | |
pyrocatechol, catechol, 1,2-dihydroxybenzene, 1,2-benzenediol, pyrocatechin, 2-hydroxyphenol, o-benzenediol, pyrocatechine, o-dihydroxybenzene, o-dioxybenzene | |
CHEBI:18135 | |
OC1=CC=CC=C1O |
Safety and Handling
GHS H Statement
Suspected of causing genetic defects.
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.<b
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cau
GHS Signal Word: Danger
EINECSNumber : 204-427-5
Recommended Storage : Product may darken during storage
RUO – Research Use Only