Please login to your online account to display your discounted pricing

Castanospermine, Tocris


Glucosidases α and β inhibitor

Quantity 50mg

Manufacturer: r & d systems  075950

 View more versions of this product

Catalog No. 075950

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 189.21
Molecular Formula C8H15NO4
CAS 79831-76-8
Chemical Name or Material Castanospermine
Assay Percent Range >98%
Synonym [1S-(1α,6β,7α,8β,8aβ)]-Octahydro-1,6,7,8-indolizinetetrol
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Castanospermine
Assay Percent Range >98%
Molecular Formula C8H15NO4
Formula Weight 189.21
Synonym [1S-(1α,6β,7α,8β,8aβ)]-Octahydro-1,6,7,8-indolizinetetrol
Accessories

Store at Room Temperature