Please login to your online account to display your discounted pricing

Castanospermine, Tocris

Glucosidases α and β inhibitor

$121.23 - $509.13

Chemical Identifiers

Chemical Name or Material Castanospermine
Assay Percent Range >98%
Molecular Formula C8H15NO4
Formula Weight 189.21
Synonym [1S-(1α,6β,7α,8β,8aβ)]-Octahydro-1,6,7,8-indolizinetetrol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

075910

 
r & d systems
075910
10mg Each for $121.23

075950

 
r & d systems
075950
50mg Each for $509.13
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 189.21
Chemical Name or Material Castanospermine
Assay Percent Range >98%
Molecular Formula C8H15NO4
CAS 79831-76-8
Synonym [1S-(1α,6β,7α,8β,8aβ)]-Octahydro-1,6,7,8-indolizinetetrol
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature