missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Epredia™ Cassette II Biopsy Tissue Cassettes
For small tissue samples, these cassettes have a front-edge lid hinge with loose-packed lids that are available in 12 different color varieties
$1,100.34 - $1,100.34
Specifications
Closure Included | Yes |
---|---|
Closure Type | Separate Lid, Front Hinge |
Description | Cassette II with Color-Matched Lids |
Packaging Type | Loose Packed Lids |
Pore Style | Small Squares |
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
B851738BE
|
Epredia™
B851738BE |
Beige |
Case of 1000 Each for $1,100.34
|
|
B851738BL
|
Epredia™
B851738BL |
Blue |
Case of 1000 Each for $1,100.34
|
|
B851738GN
|
Epredia™
B851738GN |
Green |
Case of 1000 Each for $1,100.34
|
|
B851738GY
|
Epredia™
B851738GY |
Gray |
Case of 1000 Each for $1,100.34
|
|
B851738LA
|
Epredia™
B851738LA |
Lavender |
Case of 1000 Each for $1,100.34
|
|
B851738LG
|
Epredia™
B851738LG |
Light Green |
Case of 1000 Each for $1,100.34
|
|
B851738LI
|
Epredia™
B851738LI |
Lilac |
Case of 1000 Each for $1,100.34
|
|
B851738OR
|
Epredia™
B851738OR |
Orange |
Case of 1000 Each for $1,100.34
|
|
B851738TN
|
Epredia™
B851738TN |
Tan |
Case of 1000 Each for $1,100.34
|
|
B851738WH
|
Epredia™
B851738WH |
White |
Case of 1000 Each for $1,100.34
|
|
B851738YW
|
Epredia™
B851738YW |
Yellow |
Case of 1000 Each for $1,100.34
|
|
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifications
Yes | |
Cassette II with Color-Matched Lids | |
Small Squares | |
1000/Cs |
Separate Lid, Front Hinge | |
Loose Packed Lids | |
No | |
Biopsy Cassette |