Invitrogen™ CaspGLOW™ Fluorescein Active Caspase-8 Staining Kit
Contains all the reagents necessary to detect active caspase-8 in cells with high sensitivity. Detection of the labeled cells can be determined by flow cytometry, fluorescent microscopy or a fluorescent plate reader.
Supplier: Invitrogen™ 88700542
Includes: FITC-IETD-FMK
Wash Buffer
Z-VAD-FMK
Description
- Uses the inhibitor specific for Caspase-8, Fluorescein (FITC)-conjugated IETD-FMK, for detection
- This reagent is cell permeable, non-toxic and irreversibly binds to the active enzyme
Specifications
Kit | |
Staining Kit | |
Flow Cytometry | |
Fluorescence Microscope, Flow Cytometer |
For Research Use Only