Please login to your online account to display your discounted pricing

Novus Biologicals™ Caspase Apoptosis Kit


Detect active caspases with Flow Cytometry, in vitro assay, or fluorophore-linked immunosorbent assay.

Manufacturer: novus biologicals™  NBP229383100TESTS

 View more versions of this product

 Alert

For Research Use Only

Catalog No. NBP229383

  • / Each
 

Description & Specifications

Specifications

Kit Contents 10X Wash Buffer
Quantity 100 Tests
For Use With (Application) Fluorophore-linked immunosorbent assay, Flow Cytometry, In vitro assay
Product Type Apoptosis Kit
Storage Requirements Store at 4°C. Do not freeze.

  • FLICA Apoptosis Detection kits use a novel approach to detect active caspases, using Fluorochrome Inhibitor of Caspases (FLICA).
  • Once inside the cell, the FLICA inhibitor binds covalently to the active caspase 7-11.
  • These inhibitors are cell permeable and non-cytotoxic
  • Do not use ethanol-based or methanol-based fixatives to preserve the cells – they will inactivate the FLICA label
  • Never add the fixative until the staining and final wash steps have been completed