Please login to your online account to display your discounted pricing
Novus Biologicals™ Caspase Apoptosis Kit
Detect active caspases with Flow Cytometry, in vitro assay, or fluorophore-linked immunosorbent assay.
$300.00 - $784.00
Products
Catalog Number Mfr. No. Quantity Price Quantity    
NBP229383
novus biologicals™
NBP2-29383-100Tests
100 Tests Each for $784.00
NBP22938325
novus biologicals™
NBP2-29383-25Tests
25 Tests Each for $300.00
Description & Specifications
  • FLICA Apoptosis Detection kits use a novel approach to detect active caspases, using Fluorochrome Inhibitor of Caspases (FLICA).
  • Once inside the cell, the FLICA inhibitor binds covalently to the active caspase 7-11.
  • These inhibitors are cell permeable and non-cytotoxic
  • Do not use ethanol-based or methanol-based fixatives to preserve the cells – they will inactivate the FLICA label
  • Never add the fixative until the staining and final wash steps have been completed