Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Caspase 6 (Mch 2) Ab-3, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in western blotting and immunoprecipitation procedures with Thermo Scientific™ Caspase 6 (Mch 2) Ab-3, Mouse Monoclonal Antibody.

$461.78 - $1,746.69

Specifications

Antigen Caspase 6 (Mch 2) Ab-3
Clone 6CSP03; same as 14.1.190
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1126P

 
thermo scientific™ lab vision™
MS1126P
200μg/mL 1mL Each for $1,589.53

MS1126P0

 
thermo scientific™ lab vision™
MS1126P0
200μg/mL 100μL Each for $461.78

MS1126P1

 
thermo scientific™ lab vision™
MS1126P1
200μg/mL 500μL Each for $1,112.24

MS1126P1ABX

 
thermo scientific™ lab vision™
MS1126P1ABX
1mg/mL 100μL Each for $1,214.51

MS1126PABX

 
thermo scientific™ lab vision™
MS1126PABX
1mg/mL 200μL Each for $1,746.69
Description & Specifications

Specifications

Antigen Caspase 6 (Mch 2) Ab-3
Clone 6CSP03; same as 14.1.190
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Apoptosis
Type Primary
Immunogen Recombinant human caspase 6 prodomain
Isotype IgG2a kappa

Ensure accurate, reproducible results in western blotting and immunoprecipitation procedures with Thermo Scientific™ Caspase 6 (Mch 2) Ab-3, Mouse Monoclonal Antibody.

Caspases play a major role in the transduction of the apoptotic signal and execution of apoptosis in mammalian cells. Caspase 3 (CPP32) and Caspase 6 (Mch2) are the major active caspases in apoptotic cells, and are activated in response to distinct apoptosis-inducing stimuli in all cell lines analyzed. Both CPP32 and Mch2 are present in apoptotic cells as multiple active species. Caspase-6 cleaves nuclear mitotic apparatus protein (NuMA) and mediates the shrinkage and fragmentation of nuclei.

Host Species: Mouse

Clone: 6CSP03; same as 14.1.190

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Caspase 6 prodomain

Immunogen: Recombinant human caspase 6 prodomain

Molecular Weight: 34 kDa

Positive Control: Jurkat cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO