missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Epredia™ Lab Vision™ Caspase 3 (CPP32) Ab-3 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation procedures.
Supplier: Epredia™ MS1123P0
Description
Caspase 3 is ubiquitously expressed and like other caspases is synthesized as an inactive (32kDa) proenzyme. Upon activation, Caspase 3 is cleaved at Asp28-Ser29 and Asp175-Ser176 thereby generating two subunits of 17kDa and 12kDa, respectively. Recent studies have implicated that Caspase 3 is associated with the induction of apoptosis. Activation of Caspase 3 occurs in response to a variety of apoptotic inducers including Fas-mediated apoptosis.Host Species: Mouse
Clone: 3CSP03; same as 4.1.18
Isotype: IgG2a
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: Recombinant full-length human Caspase 3 protein
Molecular Weight: 32 kDa
Positive Control: Jurkat cells, Tonsil and appendix
Cellular Localization: Predominantly Cytoplasmic with some nuclear staining
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Native verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Caspase 3 (CPP32) Ab-3 | |
Monoclonal | |
200μg/mL | |
Mouse | |
100 μL | |
Apoptosis | |
Human | |
IgG2a |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
3CSP03; same as 4.1.18 | |
Unconjugated | |
Recombinant full length human Caspase 3 protein | |
RUO | |
Primary | |
Concentrated |