Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Caspase 3 (CPP32) Ab-3, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation procedures.

Manufacturer: thermo scientific™ lab vision™  MS1123P1ABX

 View more versions of this product

Catalog No. MS1123P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Caspase 3 (CPP32) Ab-3
Quantity 100µL
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Clone 3CSP03; same as 4.1.18
Cross Reactivity Human
Format Concentrated
Immunogen Recombinant full length human Caspase 3 protein
Isotype IgG2a
Host Species Mouse
Conjugate Unlabeled
Formulation Ab without BSA/Azide (Frozen)
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Research Discipline Apoptosis
Item Description 100uL; 1mg/mL; Purified w/o BSA, azide

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation procedures with Thermo Scientific™ Caspase 3 (CPP32) Ab-3, Mouse Monoclonal Antibody.

Caspase 3 is ubiquitously expressed and like other caspases is synthesized as an inactive (32kDa) proenzyme. Upon activation, Caspase 3 is cleaved at Asp28-Ser29 and Asp175-Ser176 thereby generating two subunits of 17kDa and 12kDa, respectively. Recent studies have implicated that Caspase 3 is associated with the induction of apoptosis. Activation of Caspase 3 occurs in response to a variety of apoptotic inducers including Fas-mediated apoptosis.

Host Species: Mouse

Clone: 3CSP03; same as 4.1.18

Isotype: IgG2a

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Recombinant full-length human Caspase 3 protein

Molecular Weight: 32 kDa

Positive Control: Jurkat cells, Tonsil and appendix

Cellular Localization: Predominantly Cytoplasmic with some nuclear staining

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO