Learn More
Invitrogen™ Human CASK, GST Tag Recombinant Protein
CASK is a calcium/calmodulin-dependent serine protein kinase which is a MAGUK (membrane-associated guanylate kinase) protein family member. Recombinant human CASK (1-570) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
$794.00 - $12170.00
Specifications
Protein Length | aa 1-570 |
---|---|
Species | Human |
Recombinant | Recombinant |
Protein Tag | GST Tag |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PV6271
|
Invitrogen™
PV6271 |
10μg |
Each for $794.00
|
|
|||||
PV6272
|
Invitrogen™
PV6272 |
100μg |
Each for $2,715.00
|
|
|||||
PV6273
|
Invitrogen™
PV6273 |
1mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
CASK is a scaffold protein and the encoded protein is located at synapses in the brain. CASK associates with FG syndrome 4, mental retardation, microcephaly with pontine and cerebellar hypoplasia, and a form of X-linked mental retardation. CASK functions as a cytoskeletal membrane scaffold that coordinates signal transduction pathways within the cortical cytoskeleton.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
aa 1-570 | |
Human | |
Recombinant | |
GST Tag |