missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Casein, Hammarsten grade

$42.72 - $197.39

Specifications

CAS 9000-71-9
Synonym Casein, Hammerstein Grade
Format Powder
Sensitivity Hygroscopic
Color Off-white to Pale Yellow
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6421436 View Documents Alfa Aesar™
J6421436
500g Each for $197.39
 
AAJ6421422 View Documents Alfa Aesar™
J6421422*
100g Each for $42.72
 
Specifications

Specifications

9000-71-9
Powder
Off-white to Pale Yellow
Blocking Buffer
Casein, Hammerstein Grade
Hygroscopic
Odorless