Please login to your online account to display your discounted pricing

Alfa Aesar™ Casein Digest Standard, Certified Mass Spec Grade


Manufacturer: alfa aesar™  J67662ODL

 View more versions of this product

Catalog No. AAJ67662ODL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

For Use With (Application) For the standardizing, calibrating, and troubleshooting chromatographic separations. Particularly useful for LC-MS applications.
Odor Odorless
Solubility Soluble in water
CAS 9000-71-9
Description Yellow
Storage Requirements -30°C to -10°C
Product Type Casein Digest Standard
Format Lyophilized powder
Quantity 3 vials