Molecular Probes™ Cascade Blue™ Acetyl Azide, Trisodium Salt DFS Item
Cascade Blue™ acetyl azide, trisodium salt
Supplier:  Molecular Probes™ C2284
Catalog No. C2284
Description
Description
The amine-reactive Cascade Blue™ acetyl azide and its conjugates yield blue-fluorescence (approximate excitation/emission maxima ∼396/410 nm) that can be used as a contrasting color in multicolor applications. When compared with aminocoumarin derivatives, the sulfonated pyrene Cascade Blue™ dye shows less spectral overlap with fluorescein. In addition, Cascade Blue™ dye exhibits high absorptivity, is highly fluorescent and, unlike most dyes, resists quenching upon protein conjugation.
Specifications
Specifications
5 mg
Store in freezer (-5 to -30°C) and protect from light.
Room Temperature
Cascade Blue™
CASCADE BLUE™
396
Acetyl Azide
Amine
Active Ester, Succinimidyl Ester
Classic Dyes
410
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.