missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Casamino Acids

Supplemented to microbiological growth media.

Manufacturer:  MP Biomedicals™ 113060012

 View more versions of this product

Catalog No. MP113060012



Description

Description

Applications: Casamino acids is supplemented to microbiological growth media.
Product Description: Casamino acids are a mixture of amino acids (except tryptophan) obtained from acid hydrolysis of casein.
Specifications

Specifications

Powder
Reagents
227g
Store at Room Temperature (15-30 °C).
Documents