Carver™ Lab Presses
Range of five automatic or manual models; clamping force up to 25 tons
$6,857.94 - $7,512.40
Specifications
Normal Force 12 tons
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) Product Type Weight (English) Weight (Metric) Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Product Type Weight (English) Weight (Metric) Price Quantity  
13-872-45 Fred S Carver Inc
CFS4350
For kbr pellet pressing, glove box applications and onsite field testing, palletizing, fluid extraction, pressure forming, destructive testing and other testing procedures Standard Benchtop Press 115 lb. 52.1kg Each for $7,512.40
13-872 Fred S Carver Inc
CFS3851
For pressing oils, separating liquids from solids, filtering, plastic molding, compression and crush testing Lab Press 200 lb. 90kg Each for $6,857.94
Specifications
Description
Manual Pellet Press
  • 12-ton clamping force unit with 4 in. (10.2cm) dia. platen
  • Ideal for KBr pellet pressing, glove box applications, and onsite field testing
  • Includes integral safety shield, buffer plates, 13mm evacuable die, and counter-bored platen for positive die alignment
Self-contained, Hand-operated Press
  • 6 × 6 in. (15.2 × 15.2cm) work area and integral safety shield
  • Dual-scale gauge calibrated in metric tons and pounds
  • Adaptable for use in pressing oils, separating liquids from solids, filtering, plastic molding, compression and crush testing
Two-post 15-ton Presses
  • Available with or without heated platens
  • Self-contained, semi-automatic units with 6 × 6 in. (15.2 × 15.2cm) work areas
  • Solid construction on one base frame
  • Ten-recipe storage of pressure and time settings
  • Adjustable end-of-cycle alarm with auto opening of platens
  • Can be operated in automatic or manual modes
  • Model with heated platens can be ordered with manual cooling option
Order Info
Manual Pellet Press: Dimensions: 8L × 10.25W × 20.25 in.H (20.3 × 26 × 51.4cm). Weight: 115 lb. (52kg). Auto Pellet Press: Dimensions: 31.5L × 22.75W × 22 in.H (80 × 57.8 × 55.9cm), not including handle. Weight: 400 lb. (181kg). Self-contained, Hand-operated Press: Dimensions: 16L × 15W × 36 in.H (40.6 × 38.1 × 91.4cm). Weight: 200 lb. (91kg). Two-post 15-ton Presses: Dimensions: 20L × 35W × 40 in.H (50.8 × 88.9 × 101.6cm). Weight: without heated platens, 350 lb. (159kg); with heated platens, 400 lb. (181kg).
Accessory cage equipment, heated platens, plate and cloth equipment, and swivel bearing plates are available. Contact your Fisher Scientific Customer Service Representative or visit www.fishersci.com for details.
Description & Specifications
Specifications
12 tons