3M™ 6000 Series Cartridges
Respiratory protection for workers
Manufacturer:  3M™ 7000128250
 View more versions of this product
Catalog No. 19-070-340
Description
Description
Respiratory protection for workers
For use with 3M Facepieces 6000 Series or 3M Facepieces 7000 Series with bayonet cartridge holders against a variety of gases, vapors, and particulate hazards according to NIOSH approvals.
  • For contaminants up to 10X the Permissible Exposure Limit (PEL) with half-facepiece respirators or 50X the PEL (when quantitatively fit tested) with full-facepiece respirators or according to specific OSHA standards or applicable government regulations, whichever is lower
  • Not for use in atmospheres that are unknown or immediately dangerous to life or death (IDLH)
  • 3M Catalog# 6001, 6002, 6003, 6004, 6005, 6006, 6009S
Do not exceed maximum use concentrations established by regulatory standards. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Specifications
Specifications
Mercury Vapor, Chlorine, or Sulfur Dioxide Gas Cartridge
Gray
60/Cs.
NIOSH
6009S
3M 6000 and 7000 series respirators