missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Carteolol Hydrochloride, TCI America™

Manufacturer:  TCI America C301550MG

 View more versions of this product

Catalog No. C301550MG



Specifications

Specifications

Carteolol Hydrochloride
278°C
51781-21-6
MFCD00941499
FYBXRCFPOTXTJF-UHFFFAOYSA-N
5-[3-(tert-butylamino)-2-hydroxypropoxy]-3,4-dihydro-1H-quinolin-2-one;hydrochloride
40127
328.84
≥98.0% (HPLC,T)
White
50mg
C16H25ClN2O3
5-[3-[(1,1-Dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-2(1H)-quinolinone Hydrochloride
CC(C)(C)NCC(COC1=CC=CC2=C1CCC(=O)N2)O.Cl
328.837
CHEBI:3438
Crystal-Powder at 20°C

Chemical Identifiers

51781-21-6
328.837
FYBXRCFPOTXTJF-UHFFFAOYSA-N
40127
5-[3-(tert-butylamino)-2-hydroxypropoxy]-3,4-dihydro-1H-quinolin-2-one;hydrochloride
C16H25ClN2O3
MFCD00941499
5-[3-[(1,1-Dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-2(1H)-quinolinone Hydrochloride
CHEBI:3438
CC(C)(C)NCC(COC1=CC=CC2=C1CCC(=O)N2)O.Cl
Safety and Handling

Safety and Handling

RTECSNumber : VC8282000

TSCA : No

Documents