Please login to your online account to display your discounted pricing

Alfa Aesar™ CART (62-76), Human, Rat


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66530EXD

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66530EXD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 0.5mg
Format Solid
Sequence Tyr-Gly-Gln-Val-Pro-Met-Cys-Asp-Ala-Gly-Glu-Gln-Cys-Ala-Val (Disulfide bridge: Cys68-Cys74)
Regulatory Status RUO
Storage Requirements -30° to -10°C
Product Type CART (62-76), human, rat
Molecular Weight 1568.75
Purification Method HPLC
Cross Reactivity Human, Rat