Please login to your online account to display your discounted pricing

Alfa Aesar™ CART (62-76), Human, Rat

For cellular and molecular biology applications

Specifications

Product Type CART (62-76), human, rat
Cross Reactivity Human, Rat
Molecular Weight 1568.75
Storage Requirements -30° to -10°C
Sequence Tyr-Gly-Gln-Val-Pro-Met-Cys-Asp-Ala-Gly-Glu-Gln-Cys-Ala-Val (Disulfide bridge: Cys68-Cys74)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66530MCR

 
alfa aesar™
J66530MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66530EXD

 
alfa aesar™
J66530EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type CART (62-76), human, rat
Cross Reactivity Human, Rat
Molecular Weight 1568.75
Storage Requirements -30° to -10°C
Sequence Tyr-Gly-Gln-Val-Pro-Met-Cys-Asp-Ala-Gly-Glu-Gln-Cys-Ala-Val (Disulfide bridge: Cys68-Cys74)
Format Solid
Regulatory Status RUO
Purification Method HPLC