Please login to your online account to display your discounted pricing

Alfa Aesar™ CART (55-76), Rat

For cellular and molecular biology applications

Specifications

Product Type CART (55-76), rat
Molecular Weight 2440.81
Storage Requirements -30° to -10°C
Sequence Ile-Pro-Ile-Tyr-Glu-Lys-Lys-Tyr-Gly-Gln-Val-Pro-Met-Cys-Asp-Ala-Gly-Glu-Gln-Cys-Ala-Val (Disulfide bridge: Cys68-Cys74)
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66304EXD

 
alfa aesar™
J66304EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66304MCR

 
alfa aesar™
J66304MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type CART (55-76), rat
Molecular Weight 2440.81
Storage Requirements -30° to -10°C
Sequence Ile-Pro-Ile-Tyr-Glu-Lys-Lys-Tyr-Gly-Gln-Val-Pro-Met-Cys-Asp-Ala-Gly-Glu-Gln-Cys-Ala-Val (Disulfide bridge: Cys68-Cys74)
Format Solid
Regulatory Status RUO
Purification Method HPLC