Please login to your online account to display your discounted pricing

Carrez II Solution, Fisher Chemical


Quantity 500mL
Packaging Poly Bottle

Manufacturer: ricca chemical company  SC9102500

 View more versions of this product

Catalog No. SC9102500

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500mL
Methods For AOAC Method 984.15
Packaging Poly Bottle
CAS Min % 6.59
CAS Min % 91.0
CAS Max % 6.85
CAS 7446-20-0
CAS Max % 95.0
ChemAlert Storage Symbol Gray
Chemical Name or Material Carrez II Solution
CAS 7732-18-5

Specifications

Chemical Name or Material Carrez II Solution
CAS 7732-18-5
CAS Min % 91.0
CAS Max % 95.0
CAS 7446-20-0
CAS Min % 6.59
CAS Max % 6.85
Accessories