Please login to your online account to display your discounted pricing

Carnauba wax, refined, No. 1, yellow, ACROS Organics™


Quantity 250g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  222382500

 View more versions of this product

Catalog No. AC222382500

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder or Flakes
Quantity 250g
Color Yellow
EINECS Number 232-399-4
TSCA TSCA
Melting Point 81° to 86°C
Heavy Metals (as Pb) 40ppm max.
Synonym Brazil wax
Packaging Plastic bottle
Saponification Value 78 to 95mg KOH/g
Chemical Name or Material Carnauba wax
MDL Number MFCD00130724
CAS 8015-86-9
Acid Value 2 to 7 (mg KOH/g)
Residue after Ignition 0.25% max.

Specifications

Chemical Name or Material Carnauba wax
CAS 8015-86-9
MDL Number MFCD00130724
Synonym Brazil wax