Carnauba wax, Alfa Aesar™
Manufacturer:  Alfa Aesar 3111730
 View more versions of this product
Catalog No. AA3111730
Specifications
Specifications
Carnauba wax
Odorless
82°C to 86°C
8015-86-9
14,1846
Flakes
0.990 to 0.999
1.4500
250g
MFCD00130724
Wax
Chemical Identifiers
8015-86-9
MFCD00130724
Wax
Safety and Handling
Safety and Handling
TSCA : TSCA
SDS Documents