Please login to your online account to display your discounted pricing

Carnauba wax, Alfa Aesar™

$40.34 - $40.34

Chemical Identifiers

Chemical Name or Material Carnauba wax
CAS 8015-86-9
MDL Number MFCD00130724
Merck Index 14,1846
Synonym Wax
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA3111730

 
alfa aesar
3111730
Each for $40.34
Description & Specifications

Specifications

Melting Point 82° to 86°C
Quantity 250g
Chemical Name or Material Carnauba wax
TSCA TSCA
Physical Form Flakes
Refractive Index 1.4500
MDL Number MFCD00130724
CAS 8015-86-9
Merck Index 14,1846
Synonym Wax
Density 0.990 to 0.999
Odor Odorless