missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Carnauba wax, Alfa Aesar™

Manufacturer:  Alfa Aesar 31117A7

 View more versions of this product

Catalog No. AA31117A7



Specifications

Specifications

Carnauba wax
Odorless
82°C to 86°C
8015-86-9
14,1846
Flakes
0.990g/mL to 0.999g/mL
1.4500
5kg
MFCD00130724
Wax

Chemical Identifiers

8015-86-9
MFCD00130724
Wax
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents