Please login to your online account to display your discounted pricing

Carminic acid, 70-90%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  346630050

 View more versions of this product

Catalog No. AC346630050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥70%
Physical Form Crystalline Powder
Formula Weight 492.39
Quantity 5g
Color Red to dark red
EINECS Number 215-023-3
TSCA TSCA
Beilstein 10, II, 776
Merck Index 15, 1843
Synonym C.I. 75470
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Chemical Name or Material Carminic acid
Name Note 70-90%
Assay Percent Range 70-90%
Molecular Formula C22H20O13
MDL Number MFCD00167028
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 1260-17-9
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material Carminic acid
Name Note 70-90%
CAS 1260-17-9
Assay ≥70%
Assay Percent Range 70-90%
Molecular Formula C22H20O13
Formula Weight 492.39
MDL Number MFCD00167028
Synonym C.I. 75470
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents