Please login to your online account to display your discounted pricing

Carmine, High Purity Biological Stain (Alum Lake of Carminic Acid), ACROS Organics™

$49.86 - $49.86

Chemical Identifiers

Chemical Name or Material Carmine
CAS 1390-65-4
Purity Grade Pure
Assay ≥42.0% (carminic acid)
MDL Number MFCD00167031
Synonym C.I. 75470
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC190201000

  SDS
thermo fisher scientific
AC190201000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC190200050

  SDS
fisher scientific acros
190200050
Glass bottle 5g Each for $49.86

AC190200250

  SDS
fisher scientific acros
190200250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥42.0% (carminic acid)
Physical Form Powder
Quantity 100g
Color Dark red to red-purple
Purity Grade Pure
Merck Index 15, 2442
Packaging Glass bottle
Synonym C.I. 75470
CAS 1390-65-4
Loss on Drying 20% max.
Infrared Spectrum Authentic
UV Spectrometry Complies
Ash 12% max.
MDL Number MFCD00167031
Chemical Name or Material Carmine