Please login to your online account to display your discounted pricing

Alfa Aesar™ Carmine, calcium/aluminum lake

$48.57 - $139.31

Chemical Identifiers

Chemical Name or Material Carmine
CAS 1390-65-4
Name Note calcium/aluminum lake
MDL Number MFCD00167028
Merck Index 14,1843
Synonym C.I. 75470
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1665906

 
alfa aesar™
A16659-06
5g Each for $48.57

AAA1665914

 
alfa aesar™
A16659-14
25g Each for $139.31
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material Carmine
Name Note calcium/aluminum lake
CAS 1390-65-4
MDL Number MFCD00167028
Merck Index 14,1843
Synonym C.I. 75470
RTECS Number FH8891000