Carl Zeiss™ Microscope Stage Micrometers
Finely divided stage micrometer for calibrating optical systems with eyepiece graticule patterns. Carl Zeiss™ Microscope Stage Micrometer includes 1mm and 1/100mm intervals.
Manufacturer:  Carl Zeiss Canada Ltd 474026
 View more versions of this product
Catalog No. 12-070-63
Includes: 5 lines with 1mm intervals in y-direction, 100 lines with 1/100mm intervals in y-direction
Description
Description
  • For calibrating optical systems with eyepiece graticule patterns
  • 5 lines with 1mm intervals in y-direction
  • 100 lines with 1/100mm intervals in y-direction
Specifications
Specifications
Transmitted light microscope, eyepiece graticule
Stage micrometer
Stage micrometer, 5 lines with 1mm intervals in y-direction
Documents