Carl Zeiss™ Immersol™ Immersion Oil
Increases the resolving power of a microscope. Zeiss Immersol™ is a transparent oil with a refractive index of 1.518. It is free of fluorescence and halogen, and is certified according to ISO 8036-1/2.
Manufacturer:  Carl Zeiss™ 0000001111809
 View more versions of this product
Catalog No. 29-909-77
Description
Description
Increases the resolving power of a microscope. Zeiss Immersol™ is a transparent oil with a refractive index of 1.518. It is free of fluorescence and halogen, and is certified according to ISO 8036-1/2.
  • Bubble-free immersion oil
  • Suitable for oil immersion objectives from Carl Zeiss
Specifications
Specifications
ISO 8036-1
1.518
500mL
Immersol 518 N
Bottle