Carl Zeiss™ Immersol™ Immersion Oil DFS Item
Increases the resolving power of a microscope. Zeiss Immersol™ is a transparent oil with a refractive index of 1.518. It is free of fluorescence and halogen, and is certified according to ISO 8036-1/2.
Specifications
Refractive Index 1.518
View More Specs
Products 1
Catalog Number Mfr. No. Certifications/Compliance Quantity Package Product Type Price Quantity  
Catalog Number Mfr. No. Certifications/Compliance Quantity Package Product Type Price Quantity  
29-909-75 DFS Item
View Documents
Carl Zeiss™
0000001111808
ISO 8036-1 250mL Bottle Immersol 518 N N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Increases the resolving power of a microscope. Zeiss Immersol™ is a transparent oil with a refractive index of 1.518. It is free of fluorescence and halogen, and is certified according to ISO 8036-1/2.
  • Bubble-free immersion oil
  • Suitable for oil immersion objectives from Carl Zeiss
Specifications
Specifications
1.518
SDS