Cargille Laboratories Refractive Index Liquid Reference Standards
Choose from seven single sets of Refractive Index liquids
Supplier: Cargille 18032C
Includes:
- Data sheet
Description
- Each single set contains the number of liquids specified in the chart
- Temperature-coefficient and dispersion-value data printed on label of each vial
- Each liquid is in a 7.4cc glass vial with plastic screw cap and glass applicator rod; packed in partitioned box
Order Info
Test materials may be ordered separately. Technical Bulletins are available on request.
†Certified Set.
‡Specially selected for required high-dispersion characteristics needed in dispersion staining, focal masking and double-variation refractometry.
Specifications
Refractive Index Standards Set AAA-1/2 | |
Refractometer | |
0.01 |
1.3 to 1.39 | |
±0.0002 | |
Data sheet, 10 standards |