Please login to your online account to display your discounted pricing

Cargille Immersion Oil


Greatly increases light-gathering power with high-power objectives

Manufacturer: r p cargille laboratories  CLI16484B-1

 View more versions of this product

Catalog No. 12-369A

  • / Each
  • / Case of 12 Each
 

Description & Specifications

Specifications

Item Description Type B Immersion Oil
Refractive Index 1.5150 ±0.0002
Quantity 1 oz.
Viscosity 1250 cSt
Certifications/Compliance ISO 8036/1
Density 0.923g/cm3