Please login to your online account to display your discounted pricing

Cargille Immersion Oil


Greatly increases light-gathering power with high-power objectives

Manufacturer: r p cargille laboratories  CLI16212C

 View more versions of this product

Catalog No. 12-371-1B

  • / Each
  • / Case of 12 Each
 

Description & Specifications

Specifications

Certifications/Compliance ISO 8036/1
Density 0.877g/cm3
Refractive Index 1.5150 ±0.0002
Quantity 4 oz.
Item Description Type FF Immersion Oil
Viscosity 170 cSt