Cargille Immersion Oil
Finest immersion oil available.
Supplier: Cargille 164844OZEMD
Description
For use with 100X oil immersion objectives. Type B is recommended for most educational microscopy; contains no PCB compounds.
- Type A: Low fluorescence, low viscosity
- Type B: Low fluorescence, high viscosity
Specifications
Immersion Oil Type B | |
ISO 8036/1 | |
1.5150 ±0.0002 |
1250 cSt | |
4 oz. |