Please login to your online account to display your discounted pricing

Cargille Immersion Oil


Greatly increases light-gathering power with high-power objectives

Manufacturer: r p cargille laboratories  CLI16485

 View more versions of this product

Catalog No. 12-370-1B

  • / Each
  • / Case of 12 Each
 

Description & Specifications

Specifications

Item Description Type NVH Immersion Oil
Certifications/Compliance ISO 8036/1
Refractive Index 1.5150 ±0.0002
Density 0.919g/cm3
For Use With (Application) High-viscosity oil that meets requirements for horizontal, inverted and inclined instruments and projection equipment
Quantity 4 oz.
Viscosity 21000 cSt