Please login to your online account to display your discounted pricing

Cardiogenol C hydrochloride, Tocris™

Induces cardiomyogenesis in ESCs

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material Cardiogenol C hydrochloride
Assay Percent Range >98%
Molecular Formula C13H16N4O2·HCl
Formula Weight 296.75
Synonym 4-(2-Hydroxyethylamino)-2-(4-methoxyphenylamino)-pyrimidine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

385110

 
r & d systems
385110
10mg Each for $201.36

385150

 
r & d systems
385150
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 296.75
Quantity 10mg
Chemical Name or Material Cardiogenol C hydrochloride
Assay Percent Range >98%
Molecular Formula C13H16N4O2·HCl
CAS 1049741-55-0
Synonym 4-(2-Hydroxyethylamino)-2-(4-methoxyphenylamino)-pyrimidine hydrochloride
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C