missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cardinal Health™ Paper Sterilization Tubing

For long or odd-shaped instruments

Manufacturer:  Cardinal Health™ T90012

 View more versions of this product

Catalog No. 29-636-0959



Description

Description

  • Fracture-resistant film
  • High-strength
  • Surgical-grade
  • Provides adequate breathability and blue required sterile barrier
  • Peelable, low-lint design
  • Dual chemical indicators identify steam or EO sterilization
  • Length: 100 ft. (30.5m)

Specifications

Specifications

Sterile
Low-lint
Long or odd shaped instrument
30.5m
6/Cs.
30.5cm
Blue
100 ft. x 12 in. (30.5m x 30.5cm)
100 ft.
Surgical
12 in.
SDS
Documents