Cardinal Health™ Paper Heat-Seal Pouches
With blue film
Manufacturer:  Cardinal Health™ 91315**
 View more versions of this product
Catalog No. 19-130-0048
Description
Description
  • Fracture-resistant film with high-strength surgical-grade paper provide adequate breathability and required sterile barrier
  • Peelable, low-lint design
  • Dual chemical indicators visually identify packages exposed to steam or EtO sterilization
Specifications
Specifications
Heat seal
Designed for quick, easy presentation of sterile items, can be used in steam or Ethylene Oxide (EO) sterilization
15 in.
Yes
Paper
15 x 13 in. (38.1 x 33cm)
Blue film
13 in.
38.1cm
33cm
Documents