Please login to your online account to display your discounted pricing
Cardinal Health™ Esteem™ Stretchy Powder-Free Nitrile Exam Gloves
Firm, nonslip grip
$22.32 - $23.86
Specifications
Certifications ASTM D 6319
Color Blue
Length (English) 9.5 in.
Length (Metric) 24.13cm
Material Nitrile
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    
19-162-182
cardinal health
8816NB
Small Pack of 150 Each for $23.86
Case of 10 Pack for $228.50
19-162-181
cardinal health
8815NB
X-Small Pack of 150 Each for $23.17
Case of 10 Pack for $221.90
19-162-184
cardinal health
8818NB
Large Pack of 150 Each for $23.86
Case of 10 Pack for $228.50
19-162-183
cardinal health
8817NB
Medium Pack of 150 Each for $23.86
Case of 10 Pack for $228.50
19-162-186
cardinal health
8819NXXB
2X-Large Pack of 130 Each for $22.32
Case of 10 Pack for $213.70
19-162-185
cardinal health
8819NB
X-Large Pack of 130 Each for $22.32
Case of 10 Pack for $213.70
Description & Specifications
Specifications
Certifications ASTM D 6319
Color Blue
Length (English) 9.5 in.
Length (Metric) 24.13cm
Material Nitrile
Sterility Non-sterile
Glove Cuff Style Beaded/Rolled
Hand Ambidextrous
Latex Free Yes
Powdered Powder-free
Tensile Strength 27
  • Unique stretchy nitrile formulation for greater comfort; contain no natural rubber
  • Puncture-resistant
  • Beaded cuff resists tearing and minimizes cuff rolldown
  • Nominal length: 9.5 in. (24cm)
  • Thickness: 4.7mil
  • Color: Blue
510K approved for medical use
Meet or exceed ASTM™ D 6319 for physical properties.