Cardinal Health™ Esteem™ Nitrile Gloves with Neu-Thera™
Combine comfort and a precision fit with excellent puncture resistance
$19.86 - $20.59
Specifications
Certifications ASTM D 6319
Coating Material Fully textured
Color Blue
Length (English) 10 in.
Length (Metric) 25.9cm
View More Specs
Products 6
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-165-500A View Documents Cardinal Health™
N88RX01T
X-Small Pack of 100 Each for $20.46
Case of 10 Pack for $195.70
19-165-500C View Documents Cardinal Health™
N88RX03T
Medium Pack of 100 Each for $20.46
Case of 10 Pack for $195.70
19-165-500B View Documents Cardinal Health™
N88RX02T
Small Pack of 100 Each for $20.46
Case of 10 Pack for $195.70
19-165-500E View Documents Cardinal Health™
N88RX05T
X-Large Pack of 90 Each for $20.59
Case of 10 Pack for $197.20
19-165-500D View Documents Cardinal Health™
N88RX04T
Large Pack of 100 Each for $19.86
Case of 10 Pack for $190.20
19-165-500F View Documents Cardinal Health™
N88RX06T
2X-Large Pack of 100 Each for $19.86
Case of 10 Pack for $190.20
Description
Description
Designed for and tested by healthcare professionals, these gloves are more than just a moisturizer—they are clinically proven to improve the health of your skin.
  • Protect hands from moisture loss
  • Fully textured with beaded cuffs
  • Powder-free, nonsterile
  • Average length: 9.76 in. (24.8cm)
  • Thickness: 5.1mil at fingertip, 3.9mil at palm, 3.1mil at cuff
  • Color: Blue
Certifications
510K approved for medical use
Compliance
Meet ASTM™ D6319 Standard
Specifications
Specifications
ASTM D 6319
Blue
25.9cm
Non-sterile
Ambidextrous
Powder-free
3.9mil
Fully textured
10 in.
Nitrile
Beaded/Rolled
Yes
24