Cardinal Health™ Bio-Shield™ Wrap
Excellent barrier against airborne bacteria
Manufacturer:  Cardinal Health™ 4065
 View more versions of this product
Catalog No. 36-100-4908
Description
Description
Excellent barrier against airborne bacteria
  • Soft, drapable wrap
  • Repels fluids
  • Virtually lint free
  • Protects wrapped contents from contamination
  • Regular Duty (1.0 oz../sq. yd.)
Specifications
Specifications
Blue
Excellent barrier against airborne bacteria, protects wrapped contents from contamination, ideal for lighter specialty trays, pitchers, small basins and huck towels
60 in.
60 in.
Bio-shield™ Regular Duty Wrap
Nonsterile
60 x 60 in. (152.4 x 152.4cm)
Wet Laid Cellulose
152.4cm
152.4cm
100 Case