Please login to your online account to display your discounted pricing

Cardamonin, Tocris

Inhibitor of NF-κB activation; anti-inflammatory

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material Cardamonin
Assay Percent Range >98%
Molecular Formula C16H14O4
Formula Weight 270.28
Synonym (2E)-1-(2, 4-Dihydroxy-6-methoxyphenyl)-3-phenyl-2-propen-1-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

250910

 
r & d systems
RDS250910
10mg Each for $201.36

250950

 
r & d systems
RDS250950
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 270.28
Quantity 10mg
Chemical Name or Material Cardamonin
Assay Percent Range >98%
Molecular Formula C16H14O4
CAS 19309-14-9
Synonym (2E)-1-(2, 4-Dihydroxy-6-methoxyphenyl)-3-phenyl-2-propen-1-one
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C