Please login to your online account to display your discounted pricing

Alfa Aesar™ Carboxypeptidase Y from yeast

Chemical Identifiers

Chemical Name or Material Carboxypeptidase Y from yeast
CAS 9046-67-7
MDL Number MFCD00130723
Synonym Serine carboxypeptidase; E.C. 3.4.16.1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ65311MCR

  SDS
alfa aesar™
J65311MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Lyophilized Powder
MDL Number MFCD00130723
CAS 9046-67-7
Synonym Serine carboxypeptidase; E.C. 3.4.16.1
Chemical Name or Material Carboxypeptidase Y from yeast
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H335-H315-H319
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage −30° to −10°C
Color White to Off-white
Quantity 1mg

Applications
Carboxypeptidase Y from yeast is used for C-terminal sequencing of proteins. Also for C-terminal modification of proteins and peptides. Due to its amidase action, it is applied to the sequence analysis of peptides having amidated COOH-terminal groups such as oxytocin and vasopressin.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.