Learn More
Carboxymethylcellulose sodium salt, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1810536
Description
Carboxymethylcellulose sodium salt is used in drilling muds, in detergents as a soil-suspending agent, in resin emulsion paints, adhesives, printing inks, textile sizes and protective colloid. It acts as a stabilizer in foods. It is also employed in pharmaceuticals as a suspending agent and excipients for tablets. It is used as viscosity modifiers to stabilize the emulsions. It is used as a lubricant in artificial tears and it is used to characterize enzyme activity from endoglucanases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCarboxymethylcellulose sodium salt is used in drilling muds, in detergents as a soil-suspending agent, in resin emulsion paints, adhesives, printing inks, textile sizes and protective colloid. It acts as a stabilizer in foods. It is also employed in pharmaceuticals as a suspending agent and excipients for tablets. It is used as viscosity modifiers to stabilize the emulsions. It is used as a lubricant in artificial tears and it is used to characterize enzyme activity from endoglucanases.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Carboxymethylcellulose sodium salt | |
>300°C | |
MFCD00081472 | |
14,1829 | |
Soluble in water. | |
[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O | |
263.20 |
9004-32-4 | |
(C12 H14 O9 R6)n | |
500 g | |
carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn | |
DPXJVFZANSGRMM-UHFFFAOYNA-N | |
sodium;2,3,4,5,6-pentahydroxyhexanal;acetate | |
23706213 |
Chemical Identifiers
9004-32-4 | |
263.20 | |
DPXJVFZANSGRMM-UHFFFAOYNA-N | |
23706213 | |
[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O |
(C12 H14 O9 R6)n | |
MFCD00081472 | |
carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn | |
sodium;2,3,4,5,6-pentahydroxyhexanal;acetate |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only