Please login to your online account to display your discounted pricing
Promega™ PFC7K HQ FLEXI VECTOR 20UG
Provide a rapid, high-fidelity way to transfer protein-coding regions between a variety of Flexi vectors without the need to resequence
$398.04 - $398.04
Products
Catalog Number Mfr. No. Price Quantity    
PRC8541
promega™
C8541
Each for $398.04
Description & Specifications